missing translation for 'onlineSavingsMsg'
Learn More

RBM4 Antibody (6F6) [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30521780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30521780 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30521780 Supplier Novus Biologicals Supplier No. NBP343027AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBM4 Monoclonal antibody specifically detects RBM4 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM4
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 6F6
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DKFZp547K0918, FLJ36727, hLark, Lark homolog, MGC75138, RBM4. LARK, RBM4ALARK, RNA binding motif protein 4, RNA-binding motif protein 4, RNA-binding motif protein 4a, RNA-binding protein 4, transcriptional coactivator CoAZ, ZCCHC21, ZCRB3A, zinc finger CCHC-type and RNA binding motif 3A
Host Species Mouse
Immunogen RNA binding motif protein 4 - (Uniprot# Q9BWF3)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5936
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.