missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM46 Monoclonal Antibody (OTI1E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504551S
This item is not returnable.
View return policy
Description
RBM46 Monoclonal antibody specifically detects RBM46 in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| RBM46 | |
| Monoclonal | |
| 1.25 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O35409, Q04609 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2346, 53320 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI1E7 | |
| Unconjugated | |
| FOLH1 | |
| cancer/testis antigen 68; CT68; EG633285; ENSMUSG00000033882; fe11d09; Gm761; LOW QUALITY PROTEIN: probable RNA-binding protein 46; probable RNA-binding protein 46; RBM46; RGD1560205; RNA binding motif protein 46; RNA-binding motif protein 46; wu:fe11d09; zgc:153695 | |
| Full length human recombinant protein of human FOLH1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction