missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RBM47 Polyclonal Antibody

Product Code. 17961181
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961181 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17961181 Supplier Invitrogen™ Supplier No. PA5144035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human CACO-2 whole cell, human A431 whole cell. ICC/IF: A431 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The RBM47 gene encodes RNA-binding protein 47, which plays an important role in RNA processing and regulation. This protein is involved in various aspects of RNA metabolism, including RNA splicing, editing, stabilization, and translation. RBM47 influences gene expression by binding to specific RNA sequences, affecting post-transcriptional modifications that are critical for cellular development and function. It has been implicated in cancer biology, where changes in RBM47 expression or function can contribute to tumor suppression and progression. RBM47 is known for its role in maintaining the fidelity of RNA editing processes, impacting how cells respond to genetic or environmental changes. Research into RBM47 focuses on its involvement in RNA-mediated regulatory pathways and its potential as a therapeutic target in diseases where RNA processing errors occur, such as certain cancers and genetic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM47
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene RBM47
Gene Accession No. A0AV96
Gene Alias 9530077J19Rik; BC61; hypothetical protein; hypothetical RNA binding protein RGD1359713; NET18; RBM47; RGD1359713; RNA binding motif protein 47; RNA-binding motif protein 47; RNA-binding protein 47; si:ch211-241e15.2; wu:fj94a03
Gene Symbols RBM47
Host Species Rabbit
Immunogen E.coli-derived human RBM47 recombinant protein (Position: H66-K317).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54502
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.