missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RBM4B Polyclonal Antibody

Product Code. 13455787
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13455787 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13455787 Supplier Invitrogen™ Supplier No. PA531272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Jurkat. Predicted reactivity: Mouse (97%), Rat (97%), Pig (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Required for the translational activation of PER1 mRNA in response to circadian clock. Binds directly to the 3'-UTR of the PER1 mRNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM4B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RBM4B
Gene Accession No. Q9BQ04
Gene Alias 4921506I22Rik; AI504630; AI506404; Lark2; Neosin; RBM30; RBM4B; RBM4L; RNA binding motif protein 4B; rna-binding; RNA-binding motif protein 30; RNA-binding motif protein 4B; RNA-binding protein 30; RNA-binding protein 4B; similar to zinc responsive protein ZD7; ZCCHC15; ZCCHC21B; ZCRB3B; zinc finger CCHC-type and RNA binding motif 3B; zinc responsive protein ZD7; zinc-responsive protein ZD7
Gene Symbols RBM4B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RBM4B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83759
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.