missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM5 Polyclonal Antibody

RBM5 Polyclonal Antibody
Brand: Invitrogen PA588039
This item is not returnable.
View return policy
Description
RBM5 Polyclonal Antibody for Western Blot
Specifications
| RBM5 | |
| Polyclonal | |
| Unconjugated | |
| RBM5 | |
| D030069N10Rik; G15; H37; LUCA15; Protein G15; Putative tumor suppressor LUCA15; Rbm5; Renal carcinoma antigen NY-REN-9; RMB5; RNA binding motif protein 5; RNA-binding motif protein 5; RNA-binding protein 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10181, 300996, 83486 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.55 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| B2GV05, P52756, Q91YE7 | |
| RBM5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human RBM5 (NP_0057691). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction