missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM6 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97851AF594
This item is not returnable.
View return policy
Description
RBM6 Polyclonal specifically detects RBM6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RBM6 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DEF-3, DEF3FLJ39446,3G2, DKFZp686B0877, FLJ36517, FLJ42036, g16, HLC-11, Lung cancer antigen NY-LU-12, lung cancer protooncogene 11, NY-LU-12, Protein G16, RNA binding motif protein 6, RNA-binding motif protein 6, RNA-binding protein 6, RNA-binding protein DEF-3 | |
| Produced in rabbits immunized with E. coli-derived Human RBM6 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 10180 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction