missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBMS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584516
This item is not returnable.
View return policy
Description
Immunogen sequence: QSPSWMQPQP YILQHPGAVL TPSMEHTMSL QP Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
Specifications
| RBMS1 | |
| Polyclonal | |
| Unconjugated | |
| RBMS1 | |
| 2600014B10Rik; AI255215; C2orf12; cervical cancer oncogene 4; c-myc gene single strand binding protein 2; EMBL:AAH87094.1}; HCC-4; hypothetical protein LOC526135; MSSP; MSSP1; MSSP-1; MSSP-2; MSSP-3; Rbms1; rbms1 {ECO:0000312; RNA binding motif single stranded interacting protein 1; RNA binding motif, single stranded interacting protein 1; RNA-binding motif, single-stranded-interacting protein 1; SCR2; Single-stranded DNA-binding protein MSSP-1; suppressor of cdc 2 (cdc13) with RNA binding motif 2; suppressor of CDC2 with RNA-binding motif 2; YC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29558 | |
| RBMS1 | |
| Recombinant protein corresponding to Human RBMS1. Recombinant protein control fragment (Product #RP-105208). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction