missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBMS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598385
This item is not returnable.
View return policy
Description
RBMS2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. The RBMS proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. This protein was isolated by phenotypic complementation of cdc2 and cdc13 mutants of yeast and is thought to suppress cdc2 and cdc13 mutants through the induction of translation of cdc2.
Specifications
| RBMS2 | |
| Polyclonal | |
| Unconjugated | |
| Rbms2 | |
| RBMS2; RNA binding motif single stranded interacting protein 2; RNA binding motif, single stranded interacting protein 2; RNA-binding motif, single-stranded-interacting protein 2; SCR3; suppressor of CDC2 with RNA-binding motif 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5939 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q15434 | |
| Rbms2 | |
| Recombinant Human RNA-binding motif, single-stranded-interacting protein 2 protein (1-407AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction