missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBMY1F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144865
This item is not returnable.
View return policy
Description
Immunogen sequence: WRNDRMSTRH DGYATNDGNH PSCQETRDYA Antigen sequence identity: Mouse 44%; Rat 44%.
This gene encodes a protein containing an RNA-binding motif in the N-terminus and four SRGY (serine, arginine, glycine, tyrosine) boxes in the C-terminus. Multiple copies of this gene are found in the AZFb azoospermia factor region of chromosome Y and the encoded protein is thought to be involved in spermatogenesis. Most copies of this locus are pseudogenes, although six highly similar copies have full-length ORFs and are considered functional. Four functional copies of this gene are found within inverted repeat IR2; two functional copies of this gene are found in palindrome P3, along with two copies of PTPN13-like, Y-linked.
Specifications
| RBMY1F | |
| Polyclonal | |
| Unconjugated | |
| RBMY1F | |
| RBMY1F; RBMY1J; RNA binding motif protein, Y-linked, family 1, member F; RNA-binding motif protein, Y chromosome, family 1 member F/J; Y chromosome RNA recognition motif 2; YRRM2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 159163 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15415 | |
| RBMY1F | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 203-232. Recombinant protein control fragment (Product #RP-109889). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction