missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBP4 Monoclonal Antibody (A2)

Mouse Monoclonal Antibody
€ 138.00 - € 389.00
Specifications
| Antigen | RBP4 |
|---|---|
| Clone | A2 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17826912
![]() |
Invitrogen™
MA541671 |
200 μL |
€ 389.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17634166
|
Invitrogen™
MA543709 |
20 μL |
€ 138.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBP4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This protein belongs to the lipocalin family and is the specific carrier for retinol in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post translationally and results in defective delivery and supply to the epidermal cells.Specifications
| RBP4 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02753, P27485 | |
| 397124, 5950 | |
| Recombinant protein Retinol Binding Protein 4. The antigen corresponds to amino acid range 18-201 of the target protein. | |
| Antibody |
| A2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human, Pig | |
| RBP4 | |
| MCOPCB10; Plasma retinol binding protein 4 (PRBP); Plasma retinol-binding protein; Plasma retinol-binding protein(1-176); Plasma retinol-binding protein(1-179); Plasma retinol-binding protein(1-181); Plasma retinol-binding protein(1-182); PRBP; PRO2222; RBP; Rbp4; Rbp-4; RBPA; RDCCAS; retinol; retinol binding; retinol binding protein 4; Retinol binding protein 4 interstitial; retinol binding protein 4, cellular; retinol binding protein 4, plasma; retinol-binding protein 4; retinol-binding protein 4, interstitial | |
| RBP4 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title