Learn More
RBP4 Mouse anti-Human, CoraLite 488, Clone: 1D12B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66104-100UL
This item is not returnable.
View return policy
Description
The RBP4 antibody from Proteintech is a mouse monoclonal antibody to a fusion protein of human RBP4. This antibody recognizes human antigen. The RBP4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| RBP4 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 5950 | |
| IgG2a |
| Immunofluorescence | |
| 1D12B11 | |
| Mouse | |
| Primary | |
| Human |