Learn More
RBP4 Mouse anti-Human, CoraLite 488, Clone: 1D12B11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66104-100UL
Product Code. 17571576
  • € 450.00 / 100µL
Description
The RBP4 antibody from Proteintech is a mouse monoclonal antibody to a fusion protein of human RBP4. This antibody recognizes human antigen. The RBP4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
RBP4
Monoclonal
CoraLite 488
100 μL
5950
IgG2a
Immunofluorescence
1D12B11