missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Antibody (9G7), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | RBP4/Retinol-Binding Protein 4 |
|---|---|
| Clone | 9G7 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024003
|
Novus Biologicals
NBP3-26349-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032171
|
Novus Biologicals
NBP3-26349-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBP4/Retinol-Binding Protein 4 Monoclonal antibody specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| RBP4/Retinol-Binding Protein 4 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
| A synthesized peptide derived from Human RBP4/Retinol-Binding Protein 4 [UniProt P02753] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9G7 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5950 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title