missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Antibody (RBP4/4046), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24345FR
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Monoclonal antibody specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| DyLight 680 | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
| Recombinant fragment (around aa29-148) of human RBP4/Retinol-Binding Protein 4 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 5950 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| RBP4/4046 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction