missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Rabbit anti-Mouse, Clone: 8, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90461-100UL
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 8 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
| Recombinant Mouse RBP4/Retinol-Binding Protein 4 protein | |
| 100 μg | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 5950 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction