missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP5 Antibody (2D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00083758-M05
This item is not returnable.
View return policy
Description
RBP5 Monoclonal antibody specifically detects RBP5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| RBP5 | |
| Monoclonal | |
| Unconjugated | |
| NP_113679.1 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2D1 | |
| In 1x PBS, pH 7.4 | |
| Cellular retinol-binding protein III, CRBP3, CRBPIII, CRBP-III, HRBPiso, putative cellular retinol-binding protein CRBP III, retinol binding protein 5, cellular, retinol-binding protein 5, retinol-binding protein 5, cellular | |
| RBP5 (NP_113679.1, 1 a.a. ∽ 135 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPPNLTGYYRFVSQKNMEDYLQALNISLAVRKIALLLKPDKEIEHQGNHMTVRTLSTFRNYTVQFDVGVEFEEDLRSVDGRKCQTIVTWEEEHLVCVQKGEVPNRGWRHWLEGEMLYLELTARDAVCEQVFRKVR | |
| 0.1 mg | |
| Signal Transduction | |
| 83758 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction