missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP5 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97021AF405
This item is not returnable.
View return policy
Description
RBP5 Polyclonal specifically detects RBP5 in Human samples. It is validated for ELISA.
Specifications
| RBP5 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Alexa Fluor 405 | |
| Cellular retinol-binding protein III, CRBP3, CRBPIII, CRBP-III, HRBPiso, putative cellular retinol-binding protein CRBP III, retinol binding protein 5, cellular, retinol-binding protein 5, retinol-binding protein 5, cellular | |
| Produced in rabbits immunized with purified, recombinant RBP5 (P82980; Met1-Arg135). | |
| 0.1 mL | |
| Signal Transduction | |
| 83758 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction