missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP7 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-14541-100UL
This item is not returnable.
View return policy
Description
RBP7 Polyclonal antibody apecifically detetects RBP7 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| RBP7 | |
| Polyclonal | |
| CoraLite 594 | |
| BC063013 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| RBP7 | |
| RBP7 fusion protein Ag6045 | |
| 100 μL | |
| 116362 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction