missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPMS Antibody (OTI2G10), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73835MFV610
This item is not returnable.
View return policy
Description
RBPMS Monoclonal antibody specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry
Specifications
| RBPMS | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Flow Cytometry | |
| OTI2G10 | |
| Flow Cytometry | |
| FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
| Full length human recombinant protein of human RBPMS (NP_001008712) produced in E.coli. | |
| 0.1 mL | |
| DNA Repair, Mismatch Repair | |
| 11030 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction