missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPMS Antibody (OTI3B7), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73836FR
This item is not returnable.
View return policy
Description
RBPMS Monoclonal specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| RBPMS | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RBPMS | |
| Full length recombinant protein of human RBPMS (NP_001008712) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3B7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
| Mouse | |
| Affinity Purified | |
| DNA Repair, Mismatch Repair | |
| 11030.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction