missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPMS Antibody (OTI3B7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-03905
This item is not returnable.
View return policy
Description
RBPMS Monoclonal specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| RBPMS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150, Immunohistochemistry-Frozen Reported in scientific literature (PMID: 30073365). | |
| RBPMS | |
| Full length human recombinant protein of human RBPMS (NP_001008712) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| OTI3B7 | |
| Unconjugated | |
| FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
| Mouse | |
| 24.1 kDa | |
| 0.1 mL | |
| DNA Repair, Mismatch Repair | |
| 11030 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction