missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPMS Monoclonal Antibody (OTI1C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800217S
This item is not returnable.
View return policy
Description
RBPMS Monoclonal antibody specifically detects RBPMS in Canine, Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| RBPMS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q93062 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11030, 475599 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1C11 | |
| Unconjugated | |
| Rbpms | |
| 2010300K22Rik; 2700019M19Rik; AU017537; Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; HERMES; Rbpms; RBP-MS; RGD1561067; RNA binding protein gene with multiple splicing; RNA binding protein with multiple splicing; RNA binding protein, mRNA processing factor; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS) | |
| Full length human recombinant protein of human RBPMS produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction