missing translation for 'onlineSavingsMsg'
Learn More

RBPMS Mouse anti-Human, Clone: OTI2C3, TrueMAB™, Origene

Product Code. 17507842
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17507842 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17507842 Supplier Origene Technologies Supplier No. TA800208S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBPMS Monoclonal antibody specifically detects RBPMS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RBPMS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Rbpms
Gene Accession No. Q93062
Gene Alias 2010300K22Rik; 2700019M19Rik; AU017537; Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; HERMES; Rbpms; RBP-MS; RGD1561067; RNA binding protein gene with multiple splicing; RNA binding protein with multiple splicing; RNA binding protein, mRNA processing factor; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS)
Host Species Mouse
Immunogen Full length human recombinant protein of human RBPMS produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11030
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.