Learn More
Invitrogen™ RC74 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5117481
Product Code. 17209983
  • € 510.00 / 100µg
Description
This antibody has been tested in direct-ELISA.
INTS9 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A)and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1)and U2 (RNU2)(Baillat et al., 2005).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RC74
Polyclonal
Unconjugated
INTS9
BC028953; CPSF2L; D14Ertd231e; INT9; integrator complex subunit 9; INTS9; Protein related to CPSF subunits of 74 kDa; RC74; RC-74
Rabbit
Protein G
RUO
55756
-20°C or -80°C if preferred
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
5 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q9NV88
INTS9
Recombinant Human Integrator complex subunit 9 protein (368-658aa).