missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCAN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541841
This item is not returnable.
View return policy
Description
Peptide sequence: PGEKYELHAG TESTPSVVVH VCESETEEEE ETKNPKQKIA QTRRPDPPTA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
DSCR1L2 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A, and may play a role during central nervous system development. Highest expression occurs in heart, skeletal muscle kidney, liver and peripheral blood leukocytes.
Specifications
| RCAN3 | |
| Polyclonal | |
| Unconjugated | |
| RCAN3 | |
| AU041093; Calcipressin 3; Calcipressin-3; Csp3; Down syndrome candidate region 1-like 2; down syndrome candidate region 1-like protein 2; Down syndrome critical region gene 1-like 2; DSCR1L2; dscr1l2 protein; hRCN3; MCIP3; myocyte-enriched calcineurin-interacting protein 3; RCAN family member 3; RCAN3; RCN3; Regulator of calcineurin 3; regulator of calcineurin 3 isoform 1b,2; regulator of calcineurin family member 3; similar to Calcipressin-3 (Down syndrome candidate region 1-like protein 2) (Myocyte-enriched calcineurin interacting protein 3) (MCIP3); wu:fc68f05; zgc:113155; zgc:91820 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11123 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UKA8 | |
| RCAN3 | |
| Synthetic peptide directed towards the middle region of human RCAN3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction