missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCBTB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20116
This item is not returnable.
View return policy
Description
RCBTB1 Polyclonal specifically detects RCBTB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RCBTB1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Chronic lymphocytic leukemia deletion region gene 7 protein, CLL deletion region gene 7 protein, CLLD7MGC33184, CLLL7, E4.5, FLJ10716, GDP/GTP exchange factor (GEF)-like protein, GLP, RCC1 and BTB domain-containing protein 1, regulator of chromosome condensation (RCC1) and BTB (POZ) domain containingprotein 1, Regulator of chromosome condensation and BTB domain-containing protein 1, RP11-185C18.1 | |
| Rabbit | |
| 38 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (93%), Bovine (95%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.46mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q8NDN9 | |
| RCBTB1 | |
| Recombinant protein encompassing a sequence within the center region of human RCBTB1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 55213 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction