missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCBTB1, Mouse anti-Human, Clone: 1E4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00055213-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: VDVGKWPIFTLLSPQEIASIRKACVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLSSpecifications
| RCBTB1 | |
| Monoclonal | |
| Unconjugated | |
| regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 1 | |
| CLLD7/CLLL7/GLP/MGC33184/RP11-185C18.1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1E4 | |
| In 1x PBS, pH 7.4 | |
| NM_018191 | |
| RCBTB1 | |
| RCBTB1 (NP_060661.3, 2 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55213 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction