missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCC1 Antibody (3D11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97543AF647
This item is not returnable.
View return policy
Description
RCC1 Monoclonal specifically detects RCC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RCC1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Cell cycle regulatory protein, CHC1RCC1-I, chromosome condensation 1, Chromosome condensation protein 1, guanine nucleotide-releasing protein, regulator of chromosome condensation, regulator of chromosome condensation 1, SNHG3-RCC1, SNHG3-RCC1 readthrough transcript | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes Human, Mouse and Rat RanGEF. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 3D11 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| RCC1 | |
| Recombinant human RanGEF. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 1104 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction