Learn More
Invitrogen™ RCOR1 Monoclonal Antibody (K40046_9A7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555971
Description
Sequence of this protein is as follows: RKQKREREES EDELEEANGN NPIDIEVDQN KESKKEVPPT ETVPQVKKEK HSTQAKNRAK RKPPKGMFLS QEDVEAVSAN ATAATTVLRQ LDMELVSVKR QIQNIKQTNS ALKEKLDGGI EPYRLPEVIQ KCNARWTTEE QLLAVQAIRK YGRDFQAISD VIGNKSVVQV KNFFVNYRRR FNIDEVLQEW EAEHGKEETN GPSNQKPVKS PDNSIKMPEE
RCOR1 is a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.
Specifications
| RCOR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9UKL0 | |
| RCOR1 | |
| Recombinant human RCOR1 (immunogen range is 251-470). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K40046_9A7 | |
| Unconjugated | |
| RCOR1 | |
| 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; im:6904264; KIAA0071; LOC102554884; mKIAA0071; Protein CoREST; RCOR; Rcor1; REST corepressor 1; REST corepressor 1-like; Rocr1; si:dkey-46n18.7; zgc:158607 | |
| Mouse | |
| Protein A | |
| RUO | |
| 23186 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.