Learn More
Invitrogen™ RCOR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588800
Description
Immunogen sequence: EREESEDELE EANGNNPIDI EVDQNKESKK EVPPTETVPQ VKKEKHSTQA KNRAKRKPPK GMFLSQEDVE AVSANATAAT TVLRQLDMEL VSVKRQIQNI KQTNSALKEK LDGGIEPYRL PEVIQKCNAR WTTEEQLLAV QAIRKYGRDF QAISDVIGNK SVVQVKNFFV NYRRRFNIDE VLQEWEAEHG KEETNGPSNQ KPVKSPDNSI KMPEEEDEAP VLDVRYASAS; Positive Samples: HeLa, Jurkat, Mouse testis, Mouse brain; Cellular Location: Nucleus.
RCOR1 is a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.
Specifications
| RCOR1 | |
| Polyclonal | |
| Unconjugated | |
| RCOR1 | |
| 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; im:6904264; KIAA0071; LOC102554884; mKIAA0071; Protein CoREST; RCOR; Rcor1; REST corepressor 1; REST corepressor 1-like; Rocr1; si:dkey-46n18.7; zgc:158607 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 217864, 23186 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8CFE3, Q9UKL0 | |
| RCOR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 256-485 of human CoREST/CoREST/RCOR1 (NP_0559712). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.