missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCVRN, Mouse anti-Human, Clone: 3G10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005957-M02.50ug
This item is not returnable.
View return policy
Description
Sequence: KLEWAFSLYDVDGNGTISKNEVLEIVMAIFKMITPEDVKLLPDDENTPEKRAEKIWKYFGKNDDDKLTEKEFIEGTLANKEILRLIQFEPQKVKEKMKNSpecifications
| RCVRN | |
| Monoclonal | |
| Unconjugated | |
| recoverin | |
| RCV1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3G10 | |
| In 1x PBS, pH 7.4 | |
| NM_002903.1 | |
| RCVRN | |
| RCV1 (NP_002894.1, 101 a.a. ∼ 199 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5957 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction