missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RDBP Monoclonal Antibody (E1C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527792
This item is not returnable.
View return policy
Description
Positive Control: NELF-E-transfected 293T cells Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RDBP is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.
Specifications
| RDBP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P18615 | |
| NELFE | |
| Human NELF-E. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| E1C1 | |
| Unconjugated | |
| NELFE | |
| D17H6S45; D6S45; major histocompatibility complex gene RD; negative elongation factor complex member E; negative elongation factor complex member E, Rdbp; negative elongation factor E; negative elongation factor polypeptide E; NELF E; Nelfe; NELF-E; nuclear protein; Rd; RD RNA binding protein; RD RNA-binding protein; RDBP; RDP; RNA-binding protein RD | |
| Mouse | |
| Protein G | |
| RUO | |
| 7936 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction