missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 358.00 - € 522.00
Specifications
| Antigen | RDH10 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18021576
|
Novus Biologicals
NBP2-55418 |
100 μL |
€ 522.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605607
|
Novus Biologicals
NBP2-55418-25ul |
25 μL |
€ 358.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RDH10 Polyclonal specifically detects RDH10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RDH10 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 1.1.1, EC 1.1.1.300, EC 1.1.1.n2, retinol dehydrogenase 10, retinol dehydrogenase 10 (all-trans), SDR16C4, short chain dehydrogenase/reductase family 16C, member 4 | |
| RDH10 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 157506 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LGLFSTAGVEDYCASKFGVVGFHESLSHELKAAEKDGIKTTLVCPYLVDTGMFRGCRIRKEIEPFLPPLKPDYCVKQAMKAILTDQPMICTPRLMYIVTFMKSILPFEAVVCMYRF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title