missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00145226-D01P
This item is not returnable.
View return policy
Description
RDH12 Polyclonal antibody specifically detects RDH12 in Human samples. It is validated for Western Blot
Specifications
| RDH12 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| All-trans and 9-cis retinol dehydrogenase, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.100, FLJ30273, LCA13retinol dehydrogenase 12, all-trans and 9-cis, LCA3, retinol dehydrogenase 12, retinol dehydrogenase 12 (all-trans and 9-cis), retinol dehydrogenase 12 (all-trans/9-cis/11-cis), SDR7C2, short chain dehydrogenase/reductase family 7C, member 2 | |
| RDH12 (NP_689656.1, 1 a.a. - 316 a.a.) full-length human protein. MLVTLGLLTSFFSFLYMVAPSIRKFFAGGVCRTNVQLPGKVVVITGANTGIGKETARELASRGARVYIACRDVLKGESAASEIRVDTKNSQVLVRKLDLSDTKSIRAFAEGFLAEEKQLHILINNAGVMMCPYSKTADGFETHLGVNHLGHFLLTYLLLEQLKVSAPARVVNVSSVAHHIGKIPFHDLQSEKRYSRGFAYCHSKLANVLFTRELAKRLQGTGVTTYAVHPGVVRSELVRHSSLLCLLWRLFSPFVKTAREGAQTSLHCALAEGLEPLSGKYFSDCKRTWVSPRARNNKTAERLWNVSCELLGIRWE | |
| 0.1 mg | |
| Vision | |
| 145226 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NP_689656.1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction