Learn More
RDH14 Mouse anti-Human, Clone: OTI3E1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502316
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Exhibits an oxidoreductive catalytic activity towards retinoids. Most efficient as an NADPH-dependent retinal reductase. Displays high activity toward 9-cis and all-trans-retinol. No steroid dehydrogenase activity detected.Specifications
| RDH14 | |
| Monoclonal | |
| Unconjugated | |
| RDH14 | |
| PAN2, SDR7C4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57665 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI3E1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9HBH5 | |
| RDH14 | |
| Full length human recombit protein of human RDH14 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.