missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RDH14 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15791112 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15791112 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15791112 Supplier Invitrogen™ Supplier No. PA563341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QTSIYLASSP EVEGVSGRYF GDCKEEELLP KAMDESVARK LWDISEVMVG L Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

Retinol dehydrogenase with a clear preference for NADP. Displays high activity towards 9-cis, 11-cis and all-trans-retinol. Shows a very weak activity towards 13-cis-retinol. Has no activity towards steroid. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH14
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rdh14
Gene Accession No. Q9HBH5
Gene Alias 3110030G19Rik; Alcohol dehydrogenase PAN2; alcohol dehydrogenase PAN2; expressed sequence AW215439; AW215439; Pan2; pancreas protein 2; Rdh14; Retinol dehydrogenase 14; retinol dehydrogenase 14 (all-trans and 9-cis); retinol dehydrogenase 14 (all-trans and 9-cis)9; retinol dehydrogenase 14 (all-trans/9-cis/11-cis); RGD1565196; SDR7C4; short chain dehydrogenase/reductase family 7C member 4; short chain dehydrogenase/reductase family 7C, member 4; UNQ529/PRO1072
Gene Symbols Rdh14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RDH14. Recombinant protein control fragment (Product #RP-101631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.