missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RDM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15898203
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15898203 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15898203 Supplier Invitrogen™ Supplier No. PA569799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Dog: 93%; Guinea Pig: 91%; Human: 100%; Mouse: 83%; Pig: 93%; Rabbit: 93%; Rat: 100%.

This gene encodes a protein involved in the cellular response to cisplatin, a drug commonly used in chemotherapy. The protein encoded by this gene contains two motifs: a motif found in RAD52, a protein that functions in DNA double-strand breaks and homologous recombination, and an RNA recognition motif (RRM) that is not found in RAD52. The RAD52 motif region in RAD52 is important for protein function and may be involved in DNA binding or oligomerization. Alternatively spliced transcript variants encoding different isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RDM1
Gene Accession No. Q8NG50
Gene Alias 2410008M22Rik; AW212028; DeltaN-RDM1zeta; RAD52 homolog B; RAD52 motif 1; RAD52 motif containing 1; RAD52 motif-containing protein 1; RAD52B; RAD52B homolog; Rdm1
Gene Symbols RDM1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RDM1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201299
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.