Learn More
Greiner Bio-One Reaction Tubes
Product Code. 10515191
Click to view available options
Capacity (Metric):
1.5 mL
Max. RCF:
18,000 x g (Fixed-angle rotor)
Unit Size:
Pack of 4000
Pack of 500
Product Code. 10515191
Brand: Greiner Bio-One 616201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from polypropylene for high chemical and thermal resistance. Greiner Bio-One™ Reaction Tubes withstand temperatures from –80 to +121°C.
  • Free of heavy metals
  • Disposable
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Autoclavable to 121°C  for 20 min.
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Attached
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Translucent
Material Polypropylene
Packaging 500/Pack
Bottom Shape Conical
Max. RCF 18,000 x g (Fixed-angle rotor)