missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX Ready-to-Assay™ CaS Calcium Sensor Receptor Frozen Cells

Product Code. 17385269
Encompass
Change view
Click to view available options
Quantity:
2 Vials of Cells
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17385269 2 Vials of Cells 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17385269 Supplier DiscoverX Supplier No. HTS137RTA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Ready-to-Assay™ CaS Calcium Sensor Receptor Frozen Cells measure CASR (GPCR) activation via an increase in intracellular calcium. Ready-to-Assay kits contain two vials of thaw-and-use cells.

The Ready-to-Assay™ CaS Calcium Sensor Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. cannabinoids, CP 55,940) based activation of CASR GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress CASR alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CASR , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.

Specifications

Assay Entry CaS
For Use With (Application) Frozen Cells
Quantity 2 Vials of Cells
Target Entry CASR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.