missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX Ready-to-Assay™ LPA2 Lysophospholipid Receptor Frozen Cells
Click to view available options
Quantity:
2 Vials of Cells
Unit Size:
2 ampoules
Description
The Ready-to-Assay™ LPA2 Lysophospholipid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. LPA) based activation of LPAR2 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress LPAR2 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of LPAR2 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Specifications
Specifications
| Assay Entry | LPA2 |
| For Use With (Application) | Frozen Cells |
| Primary Agonist | LPA |
| Quantity | 2 Vials of Cells |
| Target Entry | LPAR2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction