missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments Reagents for 100 tests Copper (high)

Brand:  Hanna Instruments HI93702-01


Reagents for 100 tests Copper (high)

Product Code. 10172651

  • ONLINE EXCLUSIVE PRICE
    € 114.45
    € 76.90 / Each
    Save € 37.55
    32% Off
Only null left
Estimated Shipment: 30-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Respiratory sensitiser Category 1A

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Bicinchoninic acid disodium salt

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.