missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REC8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93738-0.1ml
This item is not returnable.
View return policy
Description
REC8 Polyclonal antibody specifically detects REC8 in Human samples. It is validated for Western Blot
Specifications
| REC8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Cohesin Rec8p, cohesion rec8p, HR21spB, meiotic recombination and sister chromatid cohesion phosphoprotein of therad21p family, meiotic recombination protein REC8 homolog, meiotic recombination protein REC8-like 1, MGC950, REC8 homolog (yeast), REC8L1human homolog of rad21, S. pombe, REC8-like 1, REC8-like 1 (yeast), Rec8p, recombination and sister chromatid cohesion protein homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human REC8 (NP_005123.2). MFYYPNVLQRHTGCFATIWLAATRGSRLVKREYLRVNVVKTCEEILNYVLVRVQPPQPGLPRPRFSLYLSAQLQIGVIRVYSQQCQYLVEDIQHILERLHRAQLQIRIDMETELPSLLLPNHLAMMETLEDAPDPFFGMMSVDPRLPSPFDIPQIRHLLEAAIPERVEEIPPEVPTEPREPERIPVTVLPPEAITILEAEPIRMLEIEGERELPEVSRRELDLLIAEEEEAILLEIPRLPPPAPAEVEGIGEALGPEELRLTGWEPGALL | |
| 0.1 mL | |
| Cell Cycle and Replication, Neuroscience | |
| 9985 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction