missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RecA-1 Antibody (HIS52), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64647AF405
This item is not returnable.
View return policy
Description
RecA-1 Monoclonal specifically detects RecA-1 in Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| RecA-1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Recognizes RECA-1, a cell surface antigen which is expressed by all rat endothelial cells. | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| HIS52 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| RecA protein, RecA1, RecA2, recaLP, Recombinase A | |
| Stromal cells from rat lymph node | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction