Learn More
IKA Receiving flask 1,000mL capacity IKA
Brand:  IKA 0003743500
Receiving flask 1,000mL capacity IKA
Product Code. 11825942
  • ONLINE EXCLUSIVE PRICE
    € 250.00
    € 241.00 / Each
    Save € 9.00
    3% Off
Only null left
Estimated Shipment: 20-04-2026