missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. baumannii Outer membrane protein Omp38 Protein
A DNA sequence encoding the Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377) Outer membrane protein Omp38, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7424-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Outer membrane protein Omp38 Protein | |
Research Use Only | |
omp38 | |
Recombinant Protein | |
E. coli | |
ANAGVTVTPL LLGYTFQDSQ HNNGGKDGNL TNGPELQDDL FVGAALGIEL TPWLGFEAEY NQVKGDVDGA SAGAEYKQKQ INGNFYVTSD LITKNYDSKI KPYVLLGAGH YKYDFDGVNR GTRGTSEEGT LGNAGVGAFW RLNDALSLRT EARATYNADE EFWNYTALAG LNVVLGGHLK PAAPVVEVAP VEPTPVTPQP QELTEDLNME LRVFFDTNKS NIKDQYKPEI AKVAEKLSEY PNATARIEGH TDNTGPRKLN ERLSLARANS VKSALVNEYN VDASRLSTQG FAWDQPIADN KTKEGRAMNR RVFATITGSR TVVVQPGQEA AAPAAAQ | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
A. baumannii | |
His-SUMO | |
Tris-based buffer, 50% glycerol |