missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant B. abortus vjbR Protein

A DNA sequence encoding the Brucella abortus (strain 2308) vjbR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7642-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029990

  • € 2060.00 / 500µg


Estimated Shipment
23-10-2020
Add to basket

Specifications

Specifications

WP_002966460.1
vjbR Protein
500 μg
Not determined.
Full Length
B. abortus
His
Tris-based buffer, 50% glycerol
29595784
Greater than 90% as determined by SDS-PAGE.
Research Use Only
vjbR
Recombinant Protein
Yeast
MSLDLVHFPN YKKTFFGSSF QSDTLALLTR IRDEIGCRYV THTYRGRVGD CTKVNSADLT VLMTLPATWV ARYSSKNYFA IDPVFQEDAP YYRNDTSAIA RDLKEDADIC PAVAELLHDA EKHGLGNLFI AVSARNPKGV AGCTVFTFEV EDEDRTQFLA RMRPRLLSLA GIIHGTVCGC KDANSVASLL TPREVDCLRW AANGKTDGEI AEILSIARWT VVTYLQNAKI KLNCSNRTSA VATALSLGII DMPEVQHLV
Videos
Documents