missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine FGF 2 Protein
Click to view available options
Quantity:
1 mg
10 μg
5 μg
50 μg
Unit Size:
10µg
1mg
50µg
5µg
Specifications
Specifications
| Gene ID (Entrez) | 281161 |
| Name | FGF 2 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | FGF2 |
| Biological Activity | The recommended concentration in responsive cells is 0.1 to 2ng/ml. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Bovine |
| Species | E. coli |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction