missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Glycolipid transfer Protein

A DNA sequence encoding the Bos taurus (Bovine) Glycolipid transfer protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6098-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16010790

  • € 3405.00 / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_786993.1
Glycolipid transfer Protein
Research Use Only
GLTP
Recombinant Protein
E. coli
ALLAEHLLRP LPADKQIETG PFLEAVSHLP PFFDCLGSPV FTPIKADISG NITKIKAVYD TNPTKFRTLQ NILEVEKEMY GAEWPKVGAT LALMWLKRGL RFIQVFLQSI CDGERDENHP NLIRVNATKA YEMALKKYHG WIVQKIFQAA LYAAPYKSDF LKALSKGQNV TEEECLEKVR LFLVNYTATI DVIYEMYTRM NAELNYKV
Greater than 90% as determined by SDS-PAGE.
327678
1 mg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers