missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Pregnancy-associated glycoprotein 1

A DNA sequence encoding the Bos taurus (Bovine) Pregnancy-associated glycoprotein 1, was expressed in the hosts and tags indicated.

€ 402.00 - € 2645.00

Specifications

Accession Number NP_776836.1
Gene ID (Entrez) 281964
Name Pregnancy-associated glycoprotein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16099950
View Documents Promotion Details
enQuireBio™
QP7639-EC-10UG
10 μg
€ 402.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16089950
View Documents Promotion Details
enQuireBio™
QP7639-EC-100UG
100 μg
€ 985.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16029960
View Documents Promotion Details
enQuireBio™
QP7639-EC-500UG
500 μg
€ 2210.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16019960
View Documents Promotion Details
enQuireBio™
QP7639-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16039960
View Documents Promotion Details
enQuireBio™
QP7639-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16009960
View Documents Promotion Details
enQuireBio™
QP7639-EC-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_776836.1
Pregnancy-associated glycoprotein 1
Not determined.
Full Length of Mature Protein
Bovine
His
Tris-based buffer, 50% glycerol
281964
Research Use Only
PAG1
Recombinant Protein
E. coli
RGSNLTTHPL RNIKDLVYMG NITIGTPPQE FQVVFDTASS DLWVPSDFCT SPACSTHVRF RHLQSSTFRL TNKTFRITYG SGRMKGVVVH DTVRIGNLVS TDQPFGLSIE EYGFEGRIYD GVLGLNYPNI SFSGAIPIFD KLKNQRAISE PVFAFYLSKD EREGSVVMFG GVDHRYYEGE LNWVPLIQAG DWSVHMDRIS IERKIIACSD GCKALVDTGT SDIVGPRRLV NNIHRLIGAI PRGSEHYVPC SEVNTLPSIV FTINGINYPV PGRAYILKDD RGRCYTTFQE NRVSSSTETW YLGDVFLRLY FSVFDRGNDR IGLARAV
Greater than 88.1% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications