Learn More
enQuireBio™ Recombinant Bovine SNRPD Protein
A cDNA sequence encoding the SNRPD was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 7875.00
Specifications
Name | SNRPD Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Bovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943929
|
enQuireBio™
QP13546-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923929
|
enQuireBio™
QP13546-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933929
|
enQuireBio™
QP13546-1MG |
1 mg |
€ 7875.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SNRPD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Greater than 60% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Bovine | |
SNRPD is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |