missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine IL-12 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
Description
The Recombinant Canine IL-12 Protein is derived from CHO. The Recombinant Canine IL-12 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 403977 |
| Molecular Weight (g/mol) | 36.3 kDa (p40), 23.6 kDa (p35) |
| Name | IL-12 |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived canine IL-12 protein, (N-terminus) Canine IL-12 p40 (Ile23-Ser329) Accession &Num; Q28268 10-His tag Canine IL-12 p35 (Arg26-Ser222) (Asp63Glu,Tyr64Leu,Ile65Tyr,Pro66Ser) Accession &Num; Q28267 10-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction